Kest Technology RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
Representative image only.
Kest Technology RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
Kest Technology | Mfr: KM-RM-DU03-1| Item: 1637989
Outstanding mixing performance in critical pharma and biotechnology applications
Call For Price
Call For Price
Buy It Together
Current Item
Kest Technology RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
Kest Technology RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
1637989
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Magnetic coupled mixer
  • Robust bearing design Zr/SiC. No particle shedding, under USP <788>
  • Designed for CIP/SIP applications
  • Easy operator handling of the drive unit using proprietary Kest-Lock
  • Advanced with aseptic design and excellent mixing performance
  • Speed Sensor technology (optional) for reliable process control
  • Mixing head designed to mix to the last drop
  • Engineering and customization options
  • Compliance with higher regulatory demands
  • 1 year warranty
More About this Item
Features:
• Designed to incorporate more energy into the tank (e.g. for dispersions)
• Mixing volume capacity: Up to 8KL
• Motor painted as per RAL 7015
Reach out to us for Customization or Retrofitting
1 Year Warranty
Specifications
  • BatteryNo
  • Power (Hz)0.75
  • DescriptionRM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk