Kest Bottom Magnetic Mixer- KMS Series
Outstanding mixing performance in critical pharma and biotechnology applications
  • Magnetic coupled mixer
  • Robust bearing design Zr/SiC. No particle shedding, under USP <788>
  • Designed for CIP/SIP applications
  • Easy operator handling of the drive unit using proprietary Kest-Lock
  • Advanced with aseptic design and excellent mixing performance
  • Speed Sensor technology (optional) for reliable process control
  • Mixing head designed to mix to the last drop
  • Engineering and customization options
  • Compliance with higher regulatory demands
Features:
• Most compact design
• Mixing volume capacity: 0 to 200L
• Unique brushless DC motor. Light weight, generates less heat. No need of a frequency converter to run the motor.
• Suitable for clean room environments
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kest Technology KMS-0/3 Drive Unit, Brushless DC with SS cover; 1/Pk 1637983 Mfr # KMS-DU0/3-0
COMPARE
KMS-0/3 Drive Unit, Brushless DC with SS cover; 1/Pk
Call For Price
Kest Technology KMS-3/7 Drive Unit, Brushless DC, Gear Box and SS cover; 1/Pk 1637984 Mfr # KMS-DU3/7-2
COMPARE
KMS-3/7 Drive Unit, Brushless DC, Gear Box and SS cover; 1/Pk
Call For Price
Kest Technology KMS-7/20 Drive Unit, Brushless DC, Gear Box and SS cover; 1/Pk 1637985 Mfr # KMS-DU7/20-2
COMPARE
KMS-7/20 Drive Unit, Brushless DC, Gear Box and SS cover; 1/Pk