Kest Bottom Magnetic Mixer- KM Series
Outstanding mixing performance in critical pharma and biotechnology applications
  • Magnetic coupled mixer
  • Robust bearing design Zr/SiC. No particle shedding, under USP <788>
  • Designed for CIP/SIP applications
  • Easy operator handling of the drive unit using proprietary Kest-Lock
  • Advanced with aseptic design and excellent mixing performance
  • Speed Sensor technology (optional) for reliable process control
  • Mixing head designed to mix to the last drop
  • Engineering and customization options
  • Compliance with higher regulatory demands
Features:
• Aseptic design
• Mixing volume capacity: Up to 22KL
• AC motor painted as per RAL 7015, also available with stainless steel cover(optional)
• Also available in ATEX version
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kest Technology KM-0/3 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637955 Mfr # KM-DU0/3-1-L0
COMPARE
KM-0/3 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-3/7 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637958 Mfr # KM-DU3/7-1-L0
COMPARE
KM-3/7 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-7/20 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637962 Mfr # KM-DU7/20-1-L0
COMPARE
KM-7/20 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-20/70 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637966 Mfr # KM-DU20/70-1-L0
COMPARE
KM-20/70 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-70/120 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637969 Mfr # KM-DU70/120-1-L0
COMPARE
KM-70/120 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637972 Mfr # KM-DU120/230-1-L0
COMPARE
KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-230/600 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637976 Mfr # KM-DU230/600-1-L0
COMPARE
KM-230/600 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-600/1300 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637979 Mfr # KM-DU600/1300-1-L0
COMPARE
KM-600/1300 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Kest Technology KM-1300/2200 Drive Unit with extension, Includes AC Motor, gearbox and outer driving head; 1/Pk 1637982 Mfr # KM-DU1300/2200-1-L1
COMPARE
KM-1300/2200 Drive Unit with extension, Includes AC Motor, gearbox and outer driving head; 1/Pk
Call For Price
Showing 1- 9 of 9