Kest Technology KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Representative image only.
Kest Technology KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Kest Technology | Mfr: KM-DU120/230-1-L0| Item: 1637972
Outstanding mixing performance in critical pharma and biotechnology applications
Call For Price
Call For Price
Buy It Together
Current Item
Kest Technology KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
Kest Technology KM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk
1637972
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Magnetic coupled mixer
  • Robust bearing design Zr/SiC. No particle shedding, under USP <788>
  • Designed for CIP/SIP applications
  • Easy operator handling of the drive unit using proprietary Kest-Lock
  • Advanced with aseptic design and excellent mixing performance
  • Speed Sensor technology (optional) for reliable process control
  • Mixing head designed to mix to the last drop
  • Engineering and customization options
  • Compliance with higher regulatory demands
  • 1 year warranty
More About this Item
Features:
• Aseptic design
• Mixing volume capacity: Up to 22KL
• AC motor painted as per RAL 7015, also available with stainless steel cover(optional)
• Also available in ATEX version
Reach out to us for Customization or Retrofitting
1 Year Warranty
Specifications
  • BatteryNo
  • Power (Hz)0.75/0.90
  • DescriptionKM-120/230 Drive Unit, Includes AC Motor, gearbox and outer driving head; 1/Pk