Kest Technology Electro polished Mixer Head, for KM-0/3 & KMS-0/3, Stainless steel, Surface finish Ra < 0.5 um; 1/Ea
Representative image only.
Kest Technology Electro polished Mixer Head, for KM-0/3 & KMS-0/3, Stainless steel, Surface finish Ra < 0.5 um; 1/Ea
Kest Technology | Mfr: KM-MH0/3A-2-SiC-EP| Item: 1637952
Call For Price
Call For Price
Product Description
Key Features
  • 1 year warranty
1 Year Warranty
Specifications
  • MaterialEN 1.4435/ASTM 316L, Silicone carbide (SiC)
  • BatteryNo
  • DescriptionElectro polished Mixer Head, for KM-0/3 & KMS-0/3, Stainless steel, Surface finish Ra < 0.5 µm; 1/Ea