Keck Tubing Quick Disconnect Adapters

Quick tubing connection without hassle
Safely and easily connect tubing to glassware or other tubing—connect and disconnect with just a quarter twist. These unique adapters eliminate broken glass ports and prevent twisted tubing. Glassware adapters 06841-51 and -52 are sold separately.
6,000.00 - ₹14,000.00INR / Pkg of 10

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Keck Tubing Adapter, 8 mm to 10 mm Id, 10/Pk 0684150 Mfr # MAKE-UP
COMPARE
Tubing Adapter, 8 mm to 10 mm Id, 10/Pk
₹6,000.00INR / Pkg of 10
Keck Glassware Adapter, Gl-14 Threaded Port or 10 mm OD Barbed Port, 10/Pk 0684151 Mfr # MAKE-UP
COMPARE
Glassware Adapter, Gl-14 Threaded Port or 10 mm OD Barbed Port, 10/Pk
₹10,600.00INR / Pkg of 10
Keck Tubing Coupling Adapter, 10/Pk, White 0684154 Mfr # MAKE-UP
COMPARE
Tubing Coupling Adapter, 10/Pk, White
₹6,400.00INR / Pkg of 10
Keck Flask/Bulkhead Adapter, 10/Pk, White 0684170 Mfr # MAKE-UP
COMPARE
Flask/Bulkhead Adapter, 10/Pk, White
₹14,000.00INR / Pkg of 10
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat