Keck Tubing Adapter, 8 mm to 10 mm Id, 10/Pk

Representative image only.

Keck Tubing Adapter, 8 mm to 10 mm Id, 10/Pk

Keck | Mfr: MAKE-UP| Item: 0684150

Quick tubing connection without hassle

₹6,000.00INR / Pkg of 10

₹6,000.00INR / Pkg of 10
Product Description

More About this Item

Safely and easily connect tubing to glassware or other tubing—connect and disconnect with just a quarter twist. These unique adapters eliminate broken glass ports and prevent twisted tubing. Glassware adapters 06841-51 and -52 are sold separately.

Specifications
  • DescriptionTubing Adapter, 8 mm to 10 mm Id, 10/Pk
Reviews
Q&A