Keck Metal Clips
Nickel-plated steel clips have superior chemical and temperature resistance
These metal taper clips securely hold conical joints under pressure. Nickel-plated steel taper clips feature superior chemical and temperature resistance—withstand temperatures to 932°>F (500°>C).
8,800.00 - ₹9,700.00INR / Pkg of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Clamp Type
Availability Pricing
Keck Metal Standard Taper Clip, Size 14, 6/Pk 0683605 Mfr # MAKE-UP
COMPARE
Nickel-plated Steel Glassware, Standard Taper, Joint Size 14
₹8,800.00INR / Pkg of 6
Keck Metal Standard Taper Clip, Size 19, 6/Pk 0683610 Mfr # MAKE-UP
COMPARE
Nickel-plated Steel Glassware, Standard Taper, Joint Size 19
₹9,600.00INR / Pkg of 6
Keck Metal Standard Taper Clip  Size 24  6/Pk 0683615 Mfr # MAKE-UP
COMPARE
Nickel-plated Steel Glassware, Standard Taper, Joint Size 24
₹9,200.00INR / Pkg of 6
Keck Metal Standard Taper Clip, Size 29, 6/Pk 0683620 Mfr # MAKE-UP
COMPARE
Nickel-plated Steel Glassware, Standard Taper, Joint Size 29
₹9,700.00INR / Pkg of 6