Keck Metal Standard Taper Clip, Size 29, 6/Pk
Representative image only.
Keck Metal Standard Taper Clip, Size 29, 6/Pk
Keck | Mfr: MAKE-UP| Item: 0683620
Nickel-plated steel clips have superior chemical and temperature resistance
₹9,700.00INR / Pkg of 6
₹9,700.00INR / Pkg of 6
Product Description
Key Features
  • 1 year warranty
More About this Item
These metal taper clips securely hold conical joints under pressure. Nickel-plated steel taper clips feature superior chemical and temperature resistance—withstand temperatures to 932°>F (500°>C).
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Steel
  • Clamp TypeGlassware, Standard Taper, Joint Size 29
  • DescriptionMetal Standard Taper Clip, Size 29, 6/Pk