Keck Ball-Joint Clips
Secure ball and socket joints—no locking screw required. Polyoxymethylene (POM) acetal resin clips are safe to 150°C (302°F). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases or concentrated acids).
9,600.00 - ₹16,000.00INR / Pkg of 12
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Keck Ball-Joint Clip, Size 13, Violet, 12/Pk 0683810 Mfr # MAKE-UP
COMPARE
Ball-Joint Clip, Size 13, Violet, 12/Pk
₹14,800.00INR / Pkg of 12
Keck Ball-Joint Clip, Size 19, Blue, 12/Pk 0683815 Mfr # MAKE-UP
COMPARE
Ball-Joint Clip, Size 19, Blue, 12/Pk
₹15,400.00INR / Pkg of 12
Keck Ball-Joint Clip, Size 29, Burgundy, 12/Pk 0683820 Mfr # MAKE-UP
COMPARE
Ball-Joint Clip, Size 29, Burgundy, 12/Pk
₹9,600.00INR / Pkg of 12
Keck Ball-Joint Clip, Size 35, ORange , 12/Pk 0683825 Mfr # MAKE-UP
COMPARE
Ball-Joint Clip, Size 35, Orange , 12/Pk
₹16,000.00INR / Pkg of 12