Keck Ball-Joint Clip, Size 13, Violet, 12/Pk
Representative image only.
Keck Ball-Joint Clip, Size 13, Violet, 12/Pk
Keck | Mfr: MAKE-UP| Item: 0683810
₹14,800.00INR / Pkg of 12
₹14,800.00INR / Pkg of 12
Product Description
More About this Item
Secure ball and socket joints—no locking screw required. Polyoxymethylene (POM) acetal resin clips are safe to 150°C (302°F). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases or concentrated acids).
Specifications
  • Clamp MaterialPolyoxymethylene (POM) Acetal
  • Clamp TypeBall Joint, Size 13
  • ColorViolet
  • DescriptionBall-Joint Clip, Size 13, Violet, 12/Pk