Jun-Air® Oil-Lubricated Piston Air Compressors
Quiet and efficient power source
  • Piston and cylinder engineered to reduce heat development and energy loss
  • Motor housing, spring mount, and specially designed chambers reduce noise and vibration
  • Oil circulation system doesn't just lubricate—it also cools the motor
These oil-lubricated compressors are quiet enough to install at the place of use. The motor housing encloses the sound-producing parts, and there are a total of four noise reduction chambers—two on the intake line and two on the pressure line. Spring-mounted motor and rubber feet dissipate vibrations.
Jun-Air's specially formulated synthetic oil optimizes lubrication, cools the motor, extends valve life, and has a long working life. Minimized tolerance between the piston and cylinder means no piston rings are required, reducing wear and tear by inhibiting heat development and energy loss.
711,000.00 - ₹994,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Power (VAC) Tank Size (Gal)
Availability Pricing
JUN-AIR Jun-Air Oil-Lubricated Piston Air Compressor, 0.71 cfm, 120 psi, 120V 50/60 Hz 1620201 Mfr # 1104360
COMPARE
0.71 120 1.1
₹711,000.00INR / Each
JUN-AIR Jun-Air Oil-Lubricated Piston Air Compressor, 2.12 cfm, 120 psi, 4 L tank, 120V 1620203 Mfr # 1152000
COMPARE
2.12 120 1.1
₹927,000.00INR / Each
JUN-AIR Jun-Air Oil-Lubricated Piston Air Compressor, 2.12 cfm, 120 psi, 25 L tank, 120V 1620205 Mfr # 1452000
COMPARE
2.12 120 6.6
₹994,000.00INR / Each