Jenway® Micro-Volume Conductivity Probes
Jenway's micro-volume probes are designed for the measurement of solutions in small containers
  • Ideal for use in test tubes
The 4 mm (027 815) and 6 mm (027 816) shaft diameters allow for accurate readings in sample volumes as small as 150 µl and 200 µl respectively. The 4 mm probe has a 120 mm reach and the 6 mm probe has a 180 mm reach, making them ideal for use in test tubes and small vessels. Both probes are made from glass and have a 7 pin DIN connector so they can be used with any of Jenway's bench conductivity meters. Please view the datasheets for more information.
45,400.00 - ₹82,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
4 mm Diameter, 120 mm Reach, Micro-Volume, Glass Conductivity Probe
₹82,000.00INR / Each
COMPARE
6 mm Diameter, 180 mm Reach, Micro-Volume, Glass Conductivity Probe
₹45,400.00INR / Each