Jeio Tech Multiposition Stirrers
Independently set and control each row, column, or position
  • Ergonomic, lightweight, and low profile to conserve bench space
  • Multi-functional with four operating modes and a timer
These multiposition stirrers have four user-friendly operating modes that allow the selection of specific rows and columns, individual stirring of each position, or simultaneous stirring of all positions. Units are easy to read with digital VFD control monitors and LED operating lamps. The advanced microprocessor PID controls accurately control stirring speed. Top plates are stainless steel for excellent chemical resistance. Brushless DC motors are quiet and durable. Gradual start-up, built in thermal shutdown, and current limit protection device provide user safety.
286,000.00 - ₹758,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
Jeio Tech M5-51M Multi-position Stirrer, 5 position 100-240V 5146000 Mfr # AAH332415U
COMPARE
5 100 to 240
₹286,000.00INR / Each
Jeio Tech M5-52M Multi-position Stirrer, 10-position 100-240V 5146002 Mfr # AAH332515U
COMPARE
10 100 to 240
₹530,000.00INR / Each
Jeio Tech M5-53M Multi-position Stirrer, 15 position 100-240V 5146004 Mfr # AAH332615U
COMPARE
15 100 to 240
₹758,000.00INR / Each