Intelli-Mixer RM-2 Rotator, Vortex Shaker
Rotator, vortex mixer, blotter all in one
  • 34 modes of operation
  • Cold room safe
Powered by V-Spin the Intelli-Mixer RM-2 is the latest in cutting-edge science. V-spin harnesses the energy of a stepper motor, allowing for a wide variety of mixing, vibrating, and rotating cycles. Featuring 15 Mix, 12 rocking, 4 blotting and 3 custom modes all in one versitile unit.
Perform a variety of mixing in tubes 0.5 to 50 ml or simultaneously vortex up to 42 eppendorf tubes with 2 different vortex frequencies. Ensure consistant, reliable, and reproducable rocking with a speed controller that automatically alternates between RPM and shakes per minute for precise control. The ultrasonic, vibration blotting modes are specifically designed for western blot protocol. Or design your own custom combination of mixing, rotating and pausing by simply holding the rack in your hand and then rotating it in any direction and at any angle. Every move is saved in the memory for future use.
A variety of accessory attachments are avaiable to make this unit a must have in the lab.
200,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
Intelli-Mixer IMIX-01 RM-2L Large Multifunctional Vortex Shaker, 1 to 99 rpm WW-51202-20 Mfr # IMIX-01
COMPARE
6.75 5.25 16
₹200,000.00INR / Each
Intelli-Mixer IMIX-02 RM-2M Medium Multifunctional Vortex Shaker, 1 to 99 rpm WW-51202-21 Mfr # IMIX-02
COMPARE
6.75 5.25 13
₹200,000.00INR / Each
Intelli-Mixer IMIX-03 RM-2S Small Multifunctional Vortex Shaker, 1 to 99 rpm WW-51202-22 Mfr # IMIX-03
COMPARE
6.75 5.25 12
₹200,000.00INR / Each