InnoCal® NIST-Traceable Calibration for Humidity/Dew Point Instruments
Supports calibrations for all types of thermohygrometers
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Calibrates hygrometers with 5 to 95% relative humidity and dew point as low as -70°C
  • Services all major brands including Cole-Parmer, Digi-Sense, Extech, Fluke, Oakton, and Vaisala
  • NIST-traceable calibration confirms your purchase meets the manufacturer’s published specification
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (EMU), and test uncertainty ratios (TUR) at no additional costs!
EPA, USDA, ISO and GLP/GMPs standards require regular NIST-traceable calibration of all test equipment being used in critical applications. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A. The results stated in our reports relate only to the items tested or calibrated.
47,900.00 - ₹99,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Number Of Calibration Test Points Description
Availability Pricing
COMPARE
Humidity 3 humidity, 1 temperature, 5 dewpoint readings NIST-Traceable Calibration; Dewpoint ≥ -40°C, 5 points
₹47,900.00INR / Each
COMPARE
Humidity 3 humidity, 1 temperature, 7 dewpoint readings NIST-Traceable Calibration; Dewpoint to -60°C, 7 points
₹58,400.00INR / Each
COMPARE
Humidity 3 humidity, 1 temperature, 8 dewpoint readings NIST-Traceable Calibration; Dewpoint to -70°C, 8 points
₹85,200.00INR / Each
COMPARE
A2LA ISO17025 Accredited Humidity 3 humidity, 1 temperature, 5 dewpoint readings A2LA ISO17025 Accredited Calibration; Dewpoint ≥ -40°C, 5 Points
₹61,000.00INR / Each
COMPARE
A2LA ISO17025 Accredited Humidity 3 humidity, 1 temperature, 7 dewpoint readings A2LA ISO17025 Accredited Calibration; Dewpoint to -60°C, 7 Points
₹72,000.00INR / Each
COMPARE
A2LA ISO17025 Accredited Humidity 3 humidity, 1 temperature, 8 dewpoint readings A2LA ISO17025 Accredited Calibration; Dewpoint to -70°C, 8 Points
₹99,800.00INR / Each