Industrial Mixer Lift Stands
Choose from three styles – manual, winch, or air to suit your application
These new heavy-duty mixer stands are constructed from heavy gauge steel and square steel tubing to accommodate loads of up to 200 lbs. Use with any of our clamp mount mixers.
Stands allow you to easily raise or lower your mixer into drums or tanks up to 48" in diameter. Each stand includes a 12" square base plate with four 9/16" mounting holes for permanent attachment to floors or dollies.
Use the manual stand for lighter mixers. Manual stand lets you move mixer up or down in 1" increments; clevis pin holds the mixer in place. The winch stand is ideal for heavier mixers. Raise the mixer to any height; winch holds the mixer in place. Choose the air-powered stand for your heaviest mixers. Air motor holds mixer in place at any level.
194,000.00 - ₹540,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Industrial Mixer Lift Stand, Manual; 30-84\" Height
₹194,000.00INR / Each
COMPARE
Industrial Mixer Lift Stand, Winch; 42-82\" Height
₹344,000.00INR / Each
COMPARE
Industrial Portable Mixer Lift Stand, Manual; 39-57\" Height
₹307,000.00INR / Each
COMPARE
Industrial Mixer Lift Stand, Pneumatic; 42-82\" Height
₹540,000.00INR / Each