Industrial Portable Mixer Lift Stand, Manual; 39-57" Height
Representative image only.
Industrial Portable Mixer Lift Stand, Manual; 39-57" Height
| Item: 0460023
Choose from three styles – manual, winch, or air to suit your application
₹353,000.00INR / Each
₹353,000.00INR / Each
Product Description
Includes
12" square base plate with four 9/16" mounting holes
Key Features
  • 1 year warranty
More About this Item
These new heavy-duty mixer stands are constructed from heavy gauge steel and square steel tubing to accommodate loads of up to 200 lbs. Use with any of our clamp mount mixers.
Stands allow you to easily raise or lower your mixer into drums or tanks up to 48" in diameter. Each stand includes a 12" square base plate with four 9/16" mounting holes for permanent attachment to floors or dollies.
Use the manual stand for lighter mixers. Manual stand lets you move mixer up or down in 1" increments; clevis pin holds the mixer in place. The winch stand is ideal for heavier mixers. Raise the mixer to any height; winch holds the mixer in place. Choose the air-powered stand for your heaviest mixers. Air motor holds mixer in place at any level.
1 Year Warranty
Specifications
  • Max Height (mm)57
  • DescriptionIndustrial Portable Mixer Lift Stand, Manual; 39-57" Height