IKA Variable-Speed Digital Orbital Shakers
Electronically controlled motor ensures accurate shaking under heavy loads
  • Ideal swivel motion with a maximum shaking weight of 4 lb (2 kg)
  • Digital display shows speed and time settings
  • Wide range of attachment combinations makes it possible to use with almost any type of vessel
Basic Model: Low-profile, quiet shaker offers long service life even under extreme load conditions. Easy to-read LED displays speed and time. Features stepless speed adjustment from 0 to 800 rpm. Operate continuously or in time operating mode (up to 50 minutes). Audio signal sounds when shaker process is complete. Select from a wide range of attachments below.
Control Model: Backlit LCD shows speed, time, and operating mode. Activate, deactivate, or set target speed or agitation time with rotary dial. Electronic time switching clock allows for both continuous operation and timed runs up to 9 hours, 59 minutes. End-point positioning feature allows automated robot-controlled sampling. All functions can be controlled and documented with LabWorldSoft® software. Order platforms and attachments separately below.
198,601.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Speed (rpm) Power (VAC) Timer Range (min)
Availability Pricing
IKA Basic Variable-Speed Digital Orbital Shaker, model, 230V 0430300 Mfr # 0002980000
COMPARE
80 230
₹198,601.00INR / Each
IKA Control model Variable-Speed Digital Orbital Shaker, , 230V 0430304 Mfr # 0002980100
COMPARE
80 230