IKA ULTRA TURRAX Tube Disperser Sample Tubes
Choose the sample tube type and size to suit your application
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
These unique disposable hermetically sealed tubes prevent sample contamination. Each sample tube contains a stirring device, blade, or ball for mixing and homogenizing small samples. Choose from three tube styles with different mixing devices to process your particular sample type.
10,240.00 - ₹20,079.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
IKA 3699600 ULTRA TURRAX Dispersion Tube 50 mL, (10/pk) 0472096 Mfr # 0003699600
COMPARE
ULTRA TURRAX Dispersion Tube 50 mL, (10/pk)
₹10,240.00INR / Pkg of 10
IKA 3699700 ULTRA TURRAX Ball Mill Tube 50 mL, stainless steel (10/pk) 0472098 Mfr # 0003699700
COMPARE
ULTRA TURRAX Ball Mill Tube 50 mL, stainless steel (10/pk)
₹12,320.00INR / Pkg of 10
IKA 3703100 ULTRA TURRAX Dispersion Tube DT-20 Tube, (25/pk) 0473807 Mfr # 0003703100
COMPARE
ULTRA TURRAX Dispersion Tube DT-20 Tube, (25/pk)
₹18,959.00INR / Pkg of 25
IKA 3703200 ULTRA TURRAX Ball Mill Tube BMT-20 S Tube, (25/pk) 0473809 Mfr # 0003703200
COMPARE
ULTRA TURRAX Ball Mill Tube BMT-20 S Tube, (25/pk)
₹20,079.00INR / Pkg of 25