IKA 3703100 ULTRA TURRAX Dispersion Tube DT-20 Tube, (25/pk)
Representative image only.
IKA 3703100 ULTRA TURRAX Dispersion Tube DT-20 Tube, (25/pk)
IKA | Mfr: 0003703100| Item: 0473807
Choose the sample tube type and size to suit your application
₹18,959.00INR / Pkg of 25
₹18,959.00INR / Pkg of 25
Product Description
Key Features
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
More About this Item
These unique disposable hermetically sealed tubes prevent sample contamination. Each sample tube contains a stirring device, blade, or ball for mixing and homogenizing small samples. Choose from three tube styles with different mixing devices to process your particular sample type.
Dispersion Tubes contain a rotor/stator element; ideal for dispersion, homogenization, and suspensions. Process sample volumes from 2 to 15 mL.
Specifications
  • Capacity (mL)15
  • Capacity (Liters)0.015
  • DescriptionULTRA TURRAX Dispersion Tube DT-20 Tube, (25/pk)