IKA Ultra-Flat Lab Disc Stirrer with Unique Desgins
Ultra-flat design makes this ideal for use in hoods or cabinets
  • Intrinsically safe
  • Different color choices available for personalization
Stirrer utilizes an advanced magnetic coil technology with no moving parts, making the drive wear-free. Lab disc can be set up to reverse rotation direction every 30 sections to ensure thorough mixing.
31,989.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
High Speed (rpm) Max Temperature (° F) Top Plate Length (in)
Availability Pricing
IKA 3362300 Ultra-Flat Lab Disc Stirrer Sunny Side Up WW-15949-34 Mfr # 3362300
COMPARE
₹31,989.00INR / Each
IKA 3578000 Ultra-Flat Lab Disc Stirrer Orange WW-15949-35 Mfr # 3578000
COMPARE
Call For Price
IKA 3578200 Ultra-Flat Lab Disc Stirrer Green WW-15949-36 Mfr # 3578200
COMPARE
Call For Price
IKA 3578400 Ultra-Flat Lab Disc Stirrer Pink WW-15949-37 Mfr # 3578400
COMPARE
Call For Price
IKA 3578600 Ultra-Flat Lab Disc Stirrer Brown WW-15949-38 Mfr # 3578600