IKA 3578400 Ultra-Flat Lab Disc Stirrer Pink
Representative image only.
IKA 3578400 Ultra-Flat Lab Disc Stirrer Pink
IKA | Mfr: 3578400| Item: 1594937
Ultra-flat design makes this ideal for use in hoods or cabinets
Call For Price
Call For Price
Product Description
Includes
cord with plug.
Key Features
  • Intrinsically safe
  • Different color choices available for personalization
More About this Item
Stirrer utilizes an advanced magnetic coil technology with no moving parts, making the drive wear-free. Lab disc can be set up to reverse rotation direction every 30 sections to ensure thorough mixing.
Specifications
  • No. Of Stirring Positions1
  • DescriptionUltra-Flat Lab Disc Stirrer Pink