IKA T-18 Ultra Turrax Digital Homogenizer
A wide speed range for a variety of applications
  • 500-watt motor processes sample volumes from 1 to 1500 mL
The 500-Watt motor offers stable operation in a variety of media samples, and variable speed control from 3,000 to 10,000 rpm. Stainless steel dispersion elements remove quickly and clean completely for reuse. The digital unit features electronic speed control and overload protection. The rotor-stator generator uses a pump-like action to constantly draw in sample. Through a combination of shear, impact, collision, and cavitation, the generator rapidly disperses and disintegrates the material suspended in the media. Easily detached rotor-stator generators makes the exchange and use of two dispersing elements seamless. Clamp sold separately.
For a complete system, you need a homogenizer and IKA L004639 Dispersing Element (04720-84).
166,657.00 - ₹470,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
IKA 3720001 T-18 Ultra Turrax Digital Homogenizer; 115 VAC 0473721 Mfr # 3720001
COMPARE
50/60 115
₹470,000.00INR / Each
IKA 3720000 T-18 Ultra Turrax Digital Homogenizer; 230 VAC 0473722 Mfr # 0003720000
COMPARE
50/60 230
₹166,657.00INR / Each