IKA® RV 3 Basic Rotary Evaporators
Updated design features digital displays for speed and temperature with a larger 4 L bath
  • Integrated safety function lifts flask out of bath in case of power failure
  • Red warning light indicates unlocked position of vapor tube
  • Single-handed manual lift offers precise positioning of glassware
  • Low device voltage (24 V) ensures user safety
  • Adjustable immersion angle
These rotary evaporators are designed and engineered for ease of use, while maintaining the highest level of safety. Manual lift handling can be done with one hand for precise positioning of glassware and is suitable for left- or right-handed operators. A flask clamping mechanism with integrated push-off function allows for easy exchange of evaporation flasks. The transparent vertical condenser's small footprint makes efficient use of the 1500 cm² cooling surface.
The unique 4 L bath design provides fast heat up, allowing for quicker experiments. Non-exposed heating coils and integrated handles offer safe operation. Bath heats from 20 to 90°C, adding versatility by allowing use of water or oil. The DC motor provides rotation speed from 20 to 300 rpm. Each unit comes standard with a dual digital display for both bath temperature and rotational speed to provide more control and easier repetition of experiments.
The evaporators are available with or without safety-coated glassware to minimize breakage and increase safety. Safety coating resists most solvents and acids while protecting you against shattered glass and dangerous reagents due to broken glass.
1,009,000.00 - ₹1,147,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bath Type Condenser Type Glass Type
Availability Pricing
IKA RV 3 V Rotary Evaporator With Uncoated Glassware And Digital Temperature Control; 100 To 240 VAC 2871130 Mfr # 0010003491
COMPARE
Water/Oil Vertical Uncoated
₹1,075,000.00INR / Each
IKA RV 3 V-C Rotary Evaporator With Coated Glassware And Digital Temperature Control; 100 To 240 VAC 2871131 Mfr # 0010003492
COMPARE
Water Vertical Safety-coated
₹1,147,000.00INR / Each
IKA RV 3 FLEX Rotary Evaporator Without Glassware And With Digital Temperature Control; 100 To 240 VAC 2871134 Mfr # 0010003493
COMPARE
Water Vertical
₹1,009,000.00INR / Each