IKA Roller Shakers
Rollers for smooth rocking and rolling action
These rolling shakers can be used with all conventional tubes and cylindrical bottles. Excellent for mixing solid and liquid suspensions, and viscous samples. Rollers can be removed to accommodate larger bottles or for quick cleaning. Choose from either the fixed-speed basic or the adjustable-speed digital with a programmable timer from 1 minute to 99 hours. All models feature easy keypad operation.
180,075.00 - ₹212,866.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm)
Availability Pricing
IKA 4010000 Roller Shaker with 6 rolls, Basic (9 7/16"W x 4 1/2"H x 12 1/2"D ) 0430401 Mfr # 0004010000
COMPARE
30
₹180,075.00INR / Each
IKA 4011000 Roller Shaker with 6 rolls, Digital (LED) 0430405 Mfr # 0004011000
COMPARE
80
₹212,866.00INR / Each
IKA 4013000 Roller Shaker with 10 rolls, Digital 0430407 Mfr # 0004013000
COMPARE
80
₹206,460.00INR / Each