IKA RH Digital Magnetic Stirrer with heating
Innovatively designed digital magnetic stirrer with heating
  • Digital display of temperature and speed for precise monitoring
  • Soft-start of the stirring motor minimizes splashing or spraying
  • Easy operation with touch keypad
  • Strong magnetic field and wide speed range for volumes up to 15 liters
  • Hot surface warning indicator prevents burns
  • Model with white-coated top plate offers excellent chemical resistance
Bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5. Highly accurate temperature control is possible with sensor placed directly in the medium. High heat output of 600 watts and stainless-steel compound hot plate ensures rapid heating. Adjustable safety temperature from 100 to 360 °C.
83,185.00 - ₹90,558.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
IKA RH Digital Magnetic Stirrer with Heating; 1/Ea 1635219 Mfr # 5019822
COMPARE
₹83,185.00INR / Each
IKA RH Digital-White Magnetic Stirrer with Heating, Enamel Plate; 1/Ea 1635220 Mfr # 4678022
COMPARE
₹90,558.00INR / Each