IKA RH Digital Magnetic Stirrer with Heating; 1/Ea
Representative image only.
IKA RH Digital Magnetic Stirrer with Heating; 1/Ea
IKA | Mfr: 5019822| Item: 1635219
Innovatively designed digital magnetic stirrer with heating
₹83,185.00INR / Each
₹83,185.00INR / Each
Product Description
Includes
Mains cable, Plastic screwdriver, H 103 protective cover, stirring bars (20, 30, 40 mm)
Key Features
  • Digital display of temperature and speed for precise monitoring
  • Soft-start of the stirring motor minimizes splashing or spraying
  • Easy operation with touch keypad
  • Strong magnetic field and wide speed range for volumes up to 15 liters
  • Hot surface warning indicator prevents burns
  • Model with white-coated top plate offers excellent chemical resistance
  • 1 year warranty
More About this Item
Bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5. Highly accurate temperature control is possible with sensor placed directly in the medium. High heat output of 600 watts and stainless-steel compound hot plate ensures rapid heating. Adjustable safety temperature from 100 to 360 °C.