IKA RH basic Magnetic Stirrer with heating
Newly designed, economic magnetic stirrer with heating.
  • Soft-start of the stirring motor
  • Includes M 10 thread for a support rod
  • Strong magnetic field and wide speed range for volumes up to 15 liters
Bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5. Highly accurate temperature control is possible with sensor placed directly in the medium. High heat output of 600 watts and stainless-steel compound hot plate ensures rapid heating. Adjustable safety temperature from 100 to 360 °C.
61,900.00 - ₹69,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
IKA RH Basic Magnetic Stirrer with Heating; 1/Ea 1635217 Mfr # 5019722
COMPARE
₹61,900.00INR / Each
IKA RH Basic-White Magnetic Stirrer with Heating, Enamel Plate; 1/Ea 1635218 Mfr # 5029722
COMPARE
₹69,400.00INR / Each