IKA RH Basic-White Magnetic Stirrer with Heating, Enamel Plate; 1/Ea
Representative image only.
IKA RH Basic-White Magnetic Stirrer with Heating, Enamel Plate; 1/Ea
IKA | Mfr: 5029722| Item: 1635218
Newly designed, economic magnetic stirrer with heating.
₹69,400.00INR / Each
₹69,400.00INR / Each
Product Description
Includes
Mains cable, Plastic screwdriver, H 102 protective cover, stirring bars (20, 30, 40 mm)
Key Features
  • Soft-start of the stirring motor
  • Includes M 10 thread for a support rod
  • Strong magnetic field and wide speed range for volumes up to 15 liters
  • 1 year warranty
More About this Item
Bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5. Highly accurate temperature control is possible with sensor placed directly in the medium. High heat output of 600 watts and stainless-steel compound hot plate ensures rapid heating. Adjustable safety temperature from 100 to 360 °C.