IKA Multiposition RT Stirring Hot Plates
Wear-free/maintenance-free magnetic coils for consistent and silent operation
  • Stir consistently in all positions
  • Adjust speed in 10-rpm increments — speed remains constant
These multiposition stirring hot plates are designed for synchronous stirring. They feature magnetic-coil technology that provides noiseless and consistent stirring on all positions. The powerful drive is 100% wear- and maintenance-free, has no moving parts, and is suitable for continuous operation. Adjust speed in 10-rpm increments; stirrer speed remains constant even when load changes. The heating plate provides homogeneous temperature distribution throughout the unit. Temperature is adjustable up to 120°C. An easy-to-use foil keypad features a digital display, an error code display, and a reverse rotation switch for superior mixing results. The closed, compact design allows easy cleaning and protects the equipment against the penetration of liquids. Stirrers are available in five-, ten-, and fifteen-position configurations.
257,000.00 - ₹989,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
IKA 3690601 RT 5 Position Stirring Hot Plate, 115V 0467194 Mfr # 3690601
COMPARE
5 115
₹839,000.00INR / Each
IKA 3690600 RT 5 Position Stirring Hot Plate, 230V 0467195 Mfr # 3690600
COMPARE
5 230
₹257,000.00INR / Each
IKA 3691101 RT 10 Position Stirring Hot Plate, 115V 0467196 Mfr # 3691101
COMPARE
10 115
₹989,000.00INR / Each
IKA 3691100 RT 10 Position Stirring Hot Plate, 230V 0467197 Mfr # 3691100
COMPARE
10 230
₹339,000.00INR / Each
IKA 3692601 RT 15 Position Stirring Hot Plate, 115V 0467198 Mfr # 3692601
COMPARE
15 115
Call For Price
IKA 3692600 RT 15 Position Stirring Hot Plate, 230V 0467199 Mfr # 3692600
COMPARE
15 230
₹427,000.00INR / Each