IKA Multiposition RO Stirrers
Wear-free/maintenance-free magnetic coils for consistent and silent operation
These multiposition stirrers are designed for synchronous stirring. They feature magnetic-coil technology that provides noiseless and consistent stirring on all positions. The powerful drive is 100% wear- and maintenance-free, has no moving parts, and is suitable for continuous operation. Adjust speed in 10 rpm increments; stirrer speed remains constant even when load changes. An easy-to-use foil keypad features a digital display, an error code display, and a reverse rotation switch for superior mixing results. The closed, compact design allows easy cleaning and protects the equipment against the penetration of liquids. Stirrers are available in five-, ten-, and fifteen-position configurations.
152,000.00 - ₹245,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
IKA 3690500 RO 5-Position Magnetic Stirrer; 100-240V 0467191 Mfr # 0003690500
COMPARE
5 100 to 240
₹152,000.00INR / Each
IKA 3691000 RO 10-Position Magnetic Stirrer; 100-240V 0467192 Mfr # 0003691000
COMPARE
10 100 to 240
₹175,000.00INR / Each
IKA 3692500 RO 15-Position Magnetic Stirrer; 100-240V 0467193 Mfr # 0003692500
COMPARE
15 100 to 240
₹245,000.00INR / Each