Ika Mini Color Squid Stirrers with Unique Designs
  • Compact and colorful
Brighten up your lab with this magnetic mini stirrer. Ideal for mixing low-viscosity liquids. Easy-to-clean glass top provides good chemical resistance. Synthetic base is made of sturdy Hytrel®. Features 0.5-watt motor with adjustable speed. Includes 5-1/2-ft cord with two-prong plug.
Various Unique Desgns available to personalize your stirrer!
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
IKA 2669936 Color Squid Mighty Mouse stirrer 1594917 Mfr # 2669936
COMPARE
Color Squid Mighty Mouse stirrer
Call For Price
IKA 2669917 Color Squid Number One Stirrer 1594918 Mfr # 2669917
COMPARE
Color Squid Number One Stirrer
Call For Price
IKA 2669922 Color Squid Wildcat Stirrer 1594919 Mfr # 2669922
COMPARE
Color Squid Wildcat Stirrer
Call For Price
IKA 3576000 Color Squid Curly Stirrer 1594920 Mfr # 3576000
COMPARE
Color Squid Curly Stirrer
Call For Price
IKA 3576300 Color Squid Sunny Stirrer 1594921 Mfr # 3576300
COMPARE
Color Squid Sunny Stirrer
Call For Price
IKA 3576600 Color Squid Bighead Stirrer 1594922 Mfr # 3576600
COMPARE
Color Squid Bighead Stirrer
Call For Price
IKA 3576900 Color Squid Balloon Stirrer 1594923 Mfr # 3576900
COMPARE
Color Squid Balloon Stirrer
Call For Price
Showing 1- 7 of 7