IKA 2669936 Color Squid Mighty Mouse stirrer
Representative image only.
IKA 2669936 Color Squid Mighty Mouse stirrer
IKA | Mfr: 2669936| Item: 1594917
Call For Price
Call For Price
Product Description
Key Features
  • Compact and colorful
More About this Item
Brighten up your lab with this magnetic mini stirrer. Ideal for mixing low-viscosity liquids. Easy-to-clean glass top provides good chemical resistance. Synthetic base is made of sturdy Hytrel®. Features 0.5-watt motor with adjustable speed. Includes 5-1/2-ft cord with two-prong plug.
Various Unique Desgns available to personalize your stirrer!
Specifications
  • No. Of Stirring Positions1
  • DescriptionColor Squid Mighty Mouse stirrer