IKA Handheld Analytical Mill
Grind dry, brittle, fibrous, or moist materials such as soil or peat
  • Fits in your hand — operate by slowly pressing down on unit
  • Universal 1/4-hp motor pulverizes material in seconds
Rugged mill has a 420 stainless steel (SS) beater for grinding substances with a hardness rating of up to 6 Mohs — order an interchangeable blade and beater separately for enhanced versatility. Heavy-duty PTFE 80-mL chamber is included — order a 250-mL chamber separately. Indicator light and safety switch protect against overheating or improper closure.
120,000.00 - ₹854,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
IKA 2900001 Economical Analytical Mill, 115 VAC, 50/60 Hz 0430010 Mfr # 2900001
COMPARE
115
₹854,000.00INR / Each
IKA 2900000 Economical Analytical Mill, 230 VAC, 50/60 Hz 0430015 Mfr # 2900000
COMPARE
230
₹120,000.00INR / Each