IKA Dry Block Heaters (Digital)
Precise temperature control for repeatable results
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes
Designed to ensure highly accurate and reproducible results, guaranteeing shorten heating times of samples. Dry heaters are ideal for determining melting and boiling points, enzyme reactions, incubation and activation of cultures, culture media tests, and in situ hybridizations.
IKA''s digital dry block heaters offer precise temperature control and rapid heat-up, and provide excellent stability to maintain results. The heaters feature a bright LCD and easy to use interface. Other features include the display of the actual temperature, user adjustable temperature set point, time display with set point, timer, counter, and external temperature probe.
Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.Modular blocks are constructed from a solid aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. (order blocks separately below)
Single block dimensions (L x W x H):
3 3⁄4" x 3" x 2" (9.5 x 7.6 x 5.1 cm).
Double block dimensions (L x W x H):
6" x 3 3⁄4" x 2 1⁄4" (15.2 x 9.5 x 5.7 cm);
double block size fits 2/4/6-block heaters.
184,863.00 - ₹431,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Number Of Blocks Power (VAC)
Availability Pricing
IKA 4025101 (115V) Digital Block Heater for Use with 1 Aluminium Blocks 115V 3670001 Mfr # 4025101
COMPARE
248 1 115
₹391,000.00INR / Each
IKA 4025201 (115V) Digital Block Heater for Use with 2 Aluminium Blocks 115V 3670003 Mfr # 4025201
COMPARE
248 2 115
₹431,000.00INR / Each
IKA 4025400 (230V) Digital Block Heater for Use with 4 Aluminium Blocks 230V 3670009 Mfr # 4025400
COMPARE
248 4 230
₹184,863.00INR / Each