IKA 4025400 (230V) Digital Block Heater for Use with 4 Aluminium Blocks 230V

Representative image only.

IKA 4025400 (230V) Digital Block Heater for Use with 4 Aluminium Blocks 230V

IKA | Mfr: 4025400| Item: 3670009

Precise temperature control for repeatable results

₹184,863.00INR / Each

₹184,863.00INR / Each
Product Description

Includes

3-wire cord with plug and PT 1000 RTD temperature probe.

Key Features

  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes
  • 2 year warranty

More About this Item

Designed to ensure highly accurate and reproducible results, guaranteeing shorten heating times of samples. Dry heaters are ideal for determining melting and boiling points, enzyme reactions, incubation and activation of cultures, culture media tests, and in situ hybridizations.

IKA''s digital dry block heaters offer precise temperature control and rapid heat-up, and provide excellent stability to maintain results. The heaters feature a bright LCD and easy to use interface. Other features include the display of the actual temperature, user adjustable temperature set point, time display with set point, timer, counter, and external temperature probe.

Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.Modular blocks are constructed from a solid aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. (order blocks separately below)

Single block dimensions (L x W x H):
3 3⁄4" x 3" x 2" (9.5 x 7.6 x 5.1 cm).
Double block dimensions (L x W x H):
6" x 3 3⁄4" x 2 1⁄4" (15.2 x 9.5 x 5.7 cm);
double block size fits 2/4/6-block heaters.

Two year warranty
Specifications
  • Number Of Blocks4
  • Max Temperature (° F)248
  • Max Temperature (° C)120
  • Power (VAC)230
  • Power (watts)375
  • Height (in)2 7/8
  • Width (in)17 61/64
  • Length (in)5 61/64
  • DescriptionDigital Block Heater for Use with 4 Aluminium Blocks 230V
  • Warranty2 year
Reviews
Q&A