IKA Dry Block Heater, Modular Heating Blocks
Precise temperature control for repeatable results
  • Modular blocks are constructed from a solid aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Single block dimensions (L x W x H):
3 3⁄4" x 3" x 2" (9.5 x 7.6 x 5.1 cm).
Double block dimensions (L x W x H):
6" x 3 3⁄4" x 2 1⁄4" (15.2 x 9.5 x 5.7 cm);
double block size fits 2/4/6-block heaters.
15,220.00 - ₹32,421.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Block Number Of Openings Block Type Description
Availability Pricing
IKA 4469300 Single Block for Use with Std Test Tube 35mm 3670032 Mfr # 0004469300
COMPARE
4 Single Block Single Block for Use with Std Test Tube 35mm
₹17,199.00INR / Each
IKA 4469700 Single Block for Use with 17 mm Vials 3670037 Mfr # 0004469700
COMPARE
12 Single Block Single Block for Use with 17 mm Vials
₹15,989.00INR / Each
IKA 4470200 Single Block for Use with 28 mm Vials 3670043 Mfr # 0004470200
COMPARE
6 Single Block Single Block for Use with 28 mm Vials
₹15,220.00INR / Each
IKA 4470600 Double Block for Use with 96 PCR Tubes 3670048 Mfr # 0004470600
COMPARE
96 Double Block Double Block for Use with 96 PCR Tubes
₹32,421.00INR / Each