Idex® VacuTight™ Chromatography Fittings with 10-32 Flat-Bottom Connection
Make dependable connections under vacuum and low pressure conditions
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
VacuTight fittings are ideal for high throughput screening and other automated liquid-handling applications due to their small size and dependable, airtight connections in low pressure and vacuum environments. The flat-bottom ferrule configuration impedes tubing inner diameter reduction and overcompression that can be a problem with many coned fittings.
1,100.00 - ₹10,500.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK WW-02014-57 Mfr # P-844X
COMPARE
1/16 PEEK VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16\" OD Tubing, 10-32 Flat Bottom; 10/PK
₹10,500.00INR / Pkg of 10
Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA WW-02017-44 Mfr # P-844
COMPARE
1/16 PEEK VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16\" OD Tubing, 10-32 Flat Bottom; 1/EA
₹1,100.00INR / Each