Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK
Representative image only.
Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK
Idex | Mfr: P-844X| Item: 0201457
Make dependable connections under vacuum and low pressure conditions
₹18,100.00INR / Pkg of 10
₹18,100.00INR / Pkg of 10
Buy It Together
Current Item
Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK
Idex VacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK
0201457
₹18,100.00INR / Pkg of 10
+ =
Total Price 32700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
VacuTight™ ferrules 02014-55.
Key Features
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
  • For 1/16" OD tubing
  • Headless, short length
  • 10/PK
  • 1 month warranty
More About this Item
VacuTight fittings are ideal for high throughput screening and other automated liquid-handling applications due to their small size and dependable, airtight connections in low pressure and vacuum environments. The flat-bottom ferrule configuration impedes tubing inner diameter reduction and overcompression that can be a problem with many coned fittings.
All VacuTight nuts must be used only with VacuTight ferrules.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorRed
  • Max Operating Pressure (PSI)800
  • Max Vacuum (in Hg)25
  • DescriptionVacuTight™ Fitting, Short, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK