Idex® Super Flangeless™ Nuts, 6-40/6-32/10-32 Flat Bottom for 1/16" OD Tubing
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,600.00 - ₹14,700.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Thread size UNF(M) (port 1) (InchShort) Description
Availability Pricing
Idex Super Flangeless™ Nut, Micro Headless, Green PEEK, 1/16" OD Tubing, 6-40 Flat Bottom; 10/PK 0201357 Mfr # M-644-03X
COMPARE
Green 6-40 flat bottom Super Flangeless™ Nut, Micro Headless, Green PEEK, 1/16" OD Tubing, 6-40 Flat Bottom; 10/PK
₹14,000.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Micro Headless, Natural PEEK, 1/16" OD Tubing, 6-32 Flat Bottom; 10/PK 0201358 Mfr # M-660X
COMPARE
Natural 6-32 flat bottom Super Flangeless™ Nut, Micro Headless, Natural PEEK, 1/16" OD Tubing, 6-32 Flat Bottom; 10/PK
₹14,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Micro Headless, Green PEEK, 1/16" OD Tubing, 6-40 Flat Bottom; 1/EA 0202126 Mfr # M-644-03
COMPARE
Green 6-40 flat bottom Super Flangeless™ Nut, Micro Headless, Green PEEK, 1/16" OD Tubing, 6-40 Flat Bottom; 1/EA
₹1,600.00INR / Each
Idex Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA 0202132 Mfr # M-653
COMPARE
Green 10-32 flat bottom Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA
₹1,600.00INR / Each
Idex Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK 0202133 Mfr # M-653X
COMPARE
Green 10-32 flat bottom Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 10/PK
₹14,600.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Micro Headless, Natural PEEK, 1/16" OD Tubing, 6-32 Flat Bottom; 1/EA 0202137 Mfr # M-660
COMPARE
Natural 6-32 flat bottom Super Flangeless™ Nut, Micro Headless, Natural PEEK, 1/16" OD Tubing, 6-32 Flat Bottom; 1/EA
₹1,600.00INR / Each