Idex Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA
Representative image only.
Idex Super Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA
Idex | Mfr: M-653| Item: 0202132
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
₹1,600.00INR / Each
₹1,600.00INR / Each
Product Description
Key Features
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 5000 psi (345 bar)
  • Headless, short
  • Green PEEK
  • 10-32 flat bottom for 1/16" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Pressure rating of nut depends on ferrule used.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)10-32 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorGreen
  • DescriptionSuper Flangeless™ Nut, Headless, Short, Green PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA