Idex® Super Flangeless™ Ferrules, 6-40 or 6-32 Flat Bottom for 1/16" OD Tubing
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
2,900.00 - ₹55,100.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Super Flangeless™ TinyTight Locking Ring, 316 SS, 1/16" OD Tubing, 6-40 Flat Bottom; 1/EA 0202125 Mfr # M-644-02
COMPARE
Natural Stainless steel Super Flangeless™ TinyTight Locking Ring, 316 SS, 1/16" OD Tubing, 6-40 Flat Bottom; 1/EA
₹2,900.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/16" OD Tubing, 6-32 or 6-40 Flat Bottom; 1/EA 0202128 Mfr # M-650
COMPARE
Natural PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/16" OD Tubing, 6-32 or 6-40 Flat Bottom; 1/EA
₹5,700.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/16" OD Tubing, 6-32 or 6-40 Flat Bottom; 10/PK 0202129 Mfr # M-650X
COMPARE
Natural PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/16" OD Tubing, 6-32 or 6-40 Flat Bottom; 10/PK
₹55,100.00INR / Pkg of 10
Idex TinyTight Ferrule for Lee 062 MINSTAC, Natural PEEK, 6-40, 1/16" OD Tubing; 1/EA 4271873 Mfr # M-647
COMPARE
Natural PEEK/Stainless Steel Ferrule for Lee 062 MINSTAC, Natural PEEK, 6-40, 1/16" OD Tubing; 1/EA
₹5,700.00INR / Each