Idex TinyTight Ferrule for Lee 062 MINSTAC, Natural PEEK, 6-40, 1/16" OD Tubing; 1/EA
Representative image only.
Idex TinyTight Ferrule for Lee 062 MINSTAC, Natural PEEK, 6-40, 1/16" OD Tubing; 1/EA
Idex | Mfr: M-647| Item: 4271873
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
₹5,700.00INR / Each
₹5,700.00INR / Each
Product Description
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Ferrule for 1/16" OD tubing and 6-40 treaded nut
  • Delivers a precise pressure range
  • Easy-to-Use
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
This TinyTight Ferrule made of PEEK and stainless steel is used with a 6-40 threaded nut and tubing with a 1/16" outer diameter. It delivers a precise pressure range, and is an easy to use alternative for Lee Company 062 MINSTAC fittings systems.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK/Stainless Steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)6-40
  • ColorNatural
  • DescriptionFerrule for Lee 062 MINSTAC, Natural PEEK, 6-40, 1/16" OD Tubing; 1/EA