Idex® Super Flangeless™ Ferrules, 5/16-24 Flat Bottom
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,500.00 - ₹13,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA 0202140 Mfr # P-140
COMPARE
3/16 ETFE/SS Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA
₹1,500.00INR / Each
Idex Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK 0202141 Mfr # P-140X
COMPARE
3/16 ETFE/SS Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK
₹13,200.00INR / Pkg of 10