Idex Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK
Representative image only.
Idex Super Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK
Idex | Mfr: P-140X| Item: 0202141
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
₹13,200.00INR / Pkg of 10
₹13,200.00INR / Pkg of 10
Product Description
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 500 psi (34 bar)
  • 10/PK
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1 Year Warranty
Specifications
  • Fitting MaterialETFE/SS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)5/16-24 flat bottom
  • Compression Size / Tube Od (Port 1) (in)3/16
  • ColorGreen
  • Max Operating Pressure (PSI)500
  • DescriptionSuper Flangeless™ Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK