Idex® Shut-Off Valves Without Fittings
Quickly stop a flow stream
  • Withstands pressures up to 500 psi!
All valves feature 1/4-28 flat-bottom ports—provide your own fittings to suit your application.
15,300.00 - ₹16,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Valve Material Volume (µL)
Availability Pricing
Idex Shut-Off Valve Without Fittings, Natural ETFE, 10.0 µL 0201699 Mfr # P-721A
COMPARE
Natural ETFE (body)/PEEK(internal rotor) 10
₹15,300.00INR / Each
Idex Shut-Off Valve Without Fittings, Blue ETFE, 2.5 µL 0201703 Mfr # P-782NF
COMPARE
Blue ETFE (body)/PEEK (internal rotor) 2.5
₹16,700.00INR / Each
Idex Shut-Off Valve Without Fittings, Blue ETFE, 10.0 µL 0201704 Mfr # P-783-01
COMPARE
Blue ETFE (body)/PEEK (internal rotor) 10
₹15,600.00INR / Each