Idex Shut-Off Valve Without Fittings, Natural ETFE, 10.0 µL
Representative image only.
Idex Shut-Off Valve Without Fittings, Natural ETFE, 10.0 µL
Idex | Mfr: P-721A| Item: 0201699
Quickly stop a flow stream
₹15,300.00INR / Each
₹15,300.00INR / Each
Product Description
Key Features
  • Withstands pressures up to 500 psi!
  • Natural ETFE/PEEK valve
  • 10.0 µL internal volume
  • 1 year warranty
More About this Item
All valves feature 1/4-28 flat-bottom ports—provide your own fittings to suit your application.
Volume listed is maximum volume with valve fully open.
1 Year Warranty
Specifications
  • Valve MaterialETFE (body)/PEEK(internal rotor)
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat-bottom
  • Volume (µL)10
  • ColorNatural
  • Max Operating Pressure (PSI)500
  • DescriptionShut-Off Valve Without Fittings, Natural ETFE, 10.0 µL; 1/EA