Idex® Short Head/Standard Knurl Plugs for 10-32 Ports
Short head is ideal for applications where space is tight
  • Easily identify columns with color coded plugs
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
330.00 - ₹4,200.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Color Material Compatible With
Availability Pricing
Idex Port Plug, Short Head/Standard Knurl, Black Acetal, 10-32 Flat-Bottom; 1/EA 0201851 Mfr # P-849
COMPARE
Black Delrin® acetal 10-32 flat-bottom port
₹4,200.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Blue Acetal, 10-32 Coned; 1/EA 0201852 Mfr # U-467B
COMPARE
Blue Delrin® acetal 10-32 coned port
₹330.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Black Acetal, 10-32 Coned; 1/EA 0201853 Mfr # U-467BLK
COMPARE
Black Delrin® acetal 10-32 coned port
₹330.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Red Acetal, 10-32 Coned; 1/EA 0201854 Mfr # U-467R
COMPARE
Red Delrin® acetal 10-32 coned port
₹330.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, White Acetal, 10-32 Coned 0201855 Mfr # U-467W
COMPARE
White Delrin® acetal 10-32 coned port
₹330.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Blue ETFE, 10-32 Coned; 1/EA 0201856 Mfr # U-468B
COMPARE
Blue ETFE 10-32 coned port
₹500.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Black ETFE, 10-32 Coned 0201857 Mfr # U-468BLK
COMPARE
Black ETFE 10-32 coned port
₹650.00INR / Each
Idex Port Plug, Short Head/Standard Knurl, Natural ETFE, 10-32 Coned 0201859 Mfr # U-468N
COMPARE
Natural ETFE 10-32 coned port
₹430.00INR / Each
Showing 1- 8 of 8