Idex Port Plug, Short Head/Standard Knurl, White Acetal, 10-32 Coned
Representative image only.
Idex Port Plug, Short Head/Standard Knurl, White Acetal, 10-32 Coned
Idex | Mfr: U-467W| Item: 0201855
Short head is ideal for applications where space is tight
₹330.00INR / Each
₹330.00INR / Each
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • For 10-32 coned port
  • White acetal construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Year Warranty
Specifications
  • MaterialDelrin® acetal
  • ColorWhite
  • Compatible With10-32 coned port
  • DescriptionPort Plug, Short Head/Standard Knurl, White Acetal, 10-32 Coned; 1/EA